1-(2-Fluoro-1,1'-biphenyl-4-yl)ethanone

Lieferant: Apollo Scientific

Synonyme: 1-(2-Fluoro-[1,1'-biphenyl]-4-yl)ethanone, 4-Acetyl-2-fluorobiphenyl

PC200463-1G
APOSPC200463-1GEA 260 EUR
APOSPC200463-1G
1-(2-Fluoro-1,1'-biphenyl-4-yl)ethanone
1-(2-Fluoro-1,1'-biphenyl-4-yl)ethanone
Formel: C₁₄H₁₁FO
Molecular Weight: 214,24 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00127698
CAS-Nummer: 42771-79-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR