Ethyl-3-cyano-4-(trifluoromethyl)pyridine-2-carboxylate
Lieferant: Apollo Scientific
PC200464-500MG
APOSPC200464-500MGEA
727
EUR
APOSPC200464-500MG
Ethyl-3-cyano-4-(trifluoromethyl)pyridine-2-carboxylate
Ethyl-3-cyano-4-(trifluoromethyl)pyridine-2-carboxylate
Formel:
C₁₀H₇F₃N₂O₂ Molecular Weight: 244,17 g/mol Schmelzpunkt: 98…99 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD28023493 CAS-Nummer: 1610047-54-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...