3-[1-(3-Fluorophenyl)-1H-1,2,3-triazol-4-yl]-1,2,4-oxadiazole-5-carbohydrazide

Lieferant: Apollo Scientific

PC200466-1G
APOSPC200466-1GEA 780 EUR
APOSPC200466-1G
3-[1-(3-Fluorophenyl)-1H-1,2,3-triazol-4-yl]-1,2,4-oxadiazole-5-carbohydrazide
3-[1-(3-Fluorophenyl)-1H-1,2,3-triazol-4-yl]-1,2,4-oxadiazole-5-carbohydrazide
Formel: C₁₁H₈FN₇O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD17780208
CAS-Nummer: 1255783-18-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR