1-(4-Chloro-2,5-difluorobenzoyl)piperidine-4-carbohydrazide

Lieferant: Apollo Scientific

PC200472-1G
APOSPC200472-1GEA 780 EUR
APOSPC200472-1G
1-(4-Chloro-2,5-difluorobenzoyl)piperidine-4-carbohydrazide
1-(4-Chloro-2,5-difluorobenzoyl)piperidine-4-carbohydrazide
Formel: C₁₃H₁₄ClF₂N₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD01843368
CAS-Nummer: 388094-06-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR