1-{[1-(3-Fluoro-4-methoxyphenyl)-1H-1,2,3-triazol-5-yl]methyl}piperazine dihydrochloride

Lieferant: Apollo Scientific

PC200480-1G
APOSPC200480-1GEA 780 EUR
APOSPC200480-1G
1-{[1-(3-Fluoro-4-methoxyphenyl)-1H-1,2,3-triazol-5-yl]methyl}piperazine dihydrochloride
1-{[1-(3-Fluoro-4-methoxyphenyl)-1H-1,2,3-triazol-5-yl]methyl}piperazine dihydrochloride
Formel: C₁₄H₂₀Cl₂FN₅O
Lagertemperatur: Raumtemperatur
MDL: MFCD28023518
CAS-Nummer: 1610376-98-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR