1-{[1-(2-Fluorophenyl)-1H-1,2,3-triazol-5-yl]methyl}piperazine dihydrochloride

Lieferant: Apollo Scientific

PC200481-1G
APOSPC200481-1GEA 780 EUR
APOSPC200481-1G
1-{[1-(2-Fluorophenyl)-1H-1,2,3-triazol-5-yl]methyl}piperazine dihydrochloride
1-{[1-(2-Fluorophenyl)-1H-1,2,3-triazol-5-yl]methyl}piperazine dihydrochloride
Formel: C₁₃H₁₈Cl₂FN₅
Lagertemperatur: Raumtemperatur
MDL: MFCD28023524
CAS-Nummer: 1338692-39-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR