Diethyl-1-[2-(4-fluorophenyl)-2-oxoethyl]-1H-1,2,3-triazole-4,5-dicarboxylate

Lieferant: Apollo Scientific

PC200482-1G
APOSPC200482-1GEA 780 EUR
APOSPC200482-1G
Diethyl-1-[2-(4-fluorophenyl)-2-oxoethyl]-1H-1,2,3-triazole-4,5-dicarboxylate
Diethyl-1-[2-(4-fluorophenyl)-2-oxoethyl]-1H-1,2,3-triazole-4,5-dicarboxylate
Formel: C₁₆H₁₆FN₃O₅
Molecular Weight: 349,32 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28023527
CAS-Nummer: 1610376-99-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR