Dimethyl-1-[2-(4-fluorophenyl)-2-oxoethyl]1H-1,2,3-triazole-4,5-dicarboxylate
Lieferant: Apollo Scientific
PC200483-1G
APOSPC200483-1GEA
780
EUR
APOSPC200483-1G
Dimethyl-1-[2-(4-fluorophenyl)-2-oxoethyl]1H-1,2,3-triazole-4,5-dicarboxylate
Dimethyl-1-[2-(4-fluorophenyl)-2-oxoethyl]1H-1,2,3-triazole-4,5-dicarboxylate
Formel:
C₁₄H₁₂FN₃O₅ Molecular Weight: 321,27 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28023528 CAS-Nummer: 1610377-03-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...