2-Methyl-6-(trifluormethyl)-4(1H)-pyrimidinon

Lieferant: Apollo Scientific

Synonyme: 4-Hydroxy-2-methyl-6-(trifluoromethyl)pyrimidine, 2-Methyl-6-(trifluoromethyl)-4-pyrimidinol

PC200489-1G
APOSPC200489-1GEA 49.3 EUR
APOSPC200489-1G
2-Methyl-6-(trifluormethyl)-4(1H)-pyrimidinon
2-Methyl-6-(trifluoromethyl)pyrimidin-4-ol
Formel: C₆H₅F₃N₂O
Molecular Weight: 178.1119 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03265321
CAS-Nummer: 2836-44-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR