3-Cyano-2-fluorobenzoic acid

Lieferant: Apollo Scientific

PC200492-1G
APOSPC200492-1GEA 74.1 EUR
APOSPC200492-1G
3-Cyano-2-fluorobenzoic acid
3-Cyano-2-fluorobenzoic acid
Formel: C₈H₄FNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09261249
CAS-Nummer: 219519-77-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR