4-Chloro-N-(2,5-difluorophenyl)benzenesulphonamide

Lieferant: Apollo Scientific

PC200495-500MG
APOSPC200495-500MGEA 661 EUR
APOSPC200495-500MG
4-Chloro-N-(2,5-difluorophenyl)benzenesulphonamide
4-Chloro-N-(2,5-difluorophenyl)benzenesulphonamide
Formel: C₁₂H₈ClF₂NO₂S
Molecular Weight: 303,72 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05136552
CAS-Nummer: 290331-05-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR