8-Brom-6-fluorchinolin

Lieferant: Apollo Scientific

PC200496-1G
APOSPC200496-1GEA 270 EUR
APOSPC200496-1G
8-Brom-6-fluorchinolin
8-Bromo-6-fluoroquinoline
Formel: C₉H₅BrFN
Molecular Weight: 226.0451 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09907837
CAS-Nummer: 22960-18-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR