8-Brom-5-fluorchinolin

Lieferant: Apollo Scientific
Achtung

PC200497-1G
APOSPC200497-1GEA 85.7 EUR
APOSPC200497-1G
8-Brom-5-fluorchinolin
8-Bromo-5-fluoroquinoline
Formel: C₉H₅BrFN
Molecular Weight: 226,0451 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09907839
CAS-Nummer: 917251-99-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR