Methyl-3-bromo-2,6-difluorobenzoate

Lieferant: Apollo Scientific

Synonyme: Methyl 3-bromo-2,6-difluorobenzoate

PC200499-500MG
APOSPC200499-500MGEA 336 EUR
APOSPC200499-500MG
Methyl-3-bromo-2,6-difluorobenzoate
Methyl-3-bromo-2,6-difluorobenzoate
Formel: C₈H₅BrF₂O₂
Molecular Weight: 251,03 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09951948
CAS-Nummer: 1378875-92-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR