2,4-Difluorophenylglyoxal hydrate 95%
Lieferant: Apollo Scientific
PC2005-5G
APOSPC2005-5GEA
263
EUR
APOSPC2005-5G
2,4-Difluorophenylglyoxal hydrate 95%
2,4-Difluorophenylglyoxal hydrate
Formel:
C₈H₆F₂O₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD04038287 CAS-Nummer: 79784-36-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...