[3-(4-Fluorophenyl)-5-methylisoxazol-4-yl]methanol 97%
Lieferant: Apollo Scientific
PC200510-250MG
APOSPC200510-250MGEA
116
EUR
APOSPC200510-250MG
[3-(4-Fluorophenyl)-5-methylisoxazol-4-yl]methanol 97%
[3-(4-Fluorophenyl)-5-methylisoxazol-4-yl]methanol
Formel:
C₁₁H₁₀FNO₂ Schmelzpunkt: 110…112 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD10008830 CAS-Nummer: 1018297-63-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...