5-(Trifluoromethyl)-1H-pyrazole-4-carboxylic acid

Lieferant: Apollo Scientific
Gefahr

PC200523-250MG
APOSPC200523-250MGEA 21.9 EUR
APOSPC200523-250MG
5-(Trifluoromethyl)-1H-pyrazole-4-carboxylic acid
5-(Trifluoromethyl)-1H-pyrazole-4-carboxylic acid
Formel: C₅H₃F₃N₂O₂
Molecular Weight: 180,08 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04125015
CAS-Nummer: 543739-84-0
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR