(3-Fluorophenoxy)acetic acid

Lieferant: Apollo Scientific

PC200534-10G PC200534-1G PC200534-5G
APOSPC200534-10GEA 248 EUR
APOSPC200534-10G APOSPC200534-1G APOSPC200534-5G
(3-Fluorophenoxy)acetic acid
(3-Fluorophenoxy)acetic acid
Formel: C₈H₇FO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD02295721
CAS-Nummer: 404-98-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR