7-Fluorochroman-4-one 97%

Lieferant: Apollo Scientific

Synonyme: 7-Fluoro-2,3-dihydro-4H-chromen-4-one

PC200546-5G
APOSPC200546-5GEA 137 EUR
APOSPC200546-5G
7-Fluorochroman-4-one 97%
7-Fluorochroman-4-one
Formel: C₉H₇FO₂
Molecular Weight: 166,15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06202344
CAS-Nummer: 113209-68-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR