7-Fluoro-4-oxo-4H-chromene-2-carboxylic acid 98%

Lieferant: Apollo Scientific

PC200548-250MG
APOSPC200548-250MGEA 79.9 EUR
APOSPC200548-250MG
7-Fluoro-4-oxo-4H-chromene-2-carboxylic acid 98%
7-Fluoro-4-oxo-4H-chromene-2-carboxylic acid
Formel: C₁₀H₅FO₄
Molecular Weight: 208,143 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11877866
CAS-Nummer: 128942-39-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR