3-(3-Fluorphenoxy)propionsäure 95+%

Lieferant: Apollo Scientific

Synonyme: 3-(3-Fluorophenoxy)propionic Acid

PC200561-1G
APOSPC200561-1GEA 69.6 EUR
APOSPC200561-1G
3-(3-Fluorphenoxy)propionsäure 95+%
3-(3-Fluorphenoxy)propionsäure
Formel: C₉H₉FO₃
Molecular Weight: 184,1644 g/mol
Schmelzpunkt: 92…96 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01931913
CAS-Nummer: 133077-42-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR