4-[3,5-Di(trifluoromethyl)phenyl]-3-thiosemicarbazide
Lieferant: Apollo Scientific
|
Achtung
|
PC2009-2G
APOSPC2009-2GEA
109
EUR
APOSPC2009-2G
4-[3,5-Di(trifluoromethyl)phenyl]-3-thiosemicarbazide
4-[3,5-Di(trifluoromethyl)phenyl]-3-thiosemicarbazide
Formel:
C₉H₇F₆N₃S Molecular Weight: 303,23 g/mol Schmelzpunkt: 158…160 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00041292 CAS-Nummer: 38901-31-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...