3-Methoxy-4-(trifluoromethyl)phenylamine 98%

Lieferant: Apollo Scientific

Synonyme: 3-Methoxy-4-(trifluoromethyl)aniline

PC201016-1G
APOSPC201016-1GEA 116 EUR
APOSPC201016-1G
3-Methoxy-4-(trifluoromethyl)phenylamine 98%
3-Methoxy-4-(trifluoromethyl)phenylamine
Formel: C₈H₈F₃NO
Molecular Weight: 191,15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11840978
CAS-Nummer: 106877-20-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR