8-(Trifluoromethyl)-1,2,3,4-tetrahydroisoquinoline hydrochloride 95+%
Lieferant: Apollo Scientific
PC201018-1G
APOSPC201018-1GEA
429
EUR
APOSPC201018-1G
8-(Trifluoromethyl)-1,2,3,4-tetrahydroisoquinoline hydrochloride 95+%
8-(Trifluoromethyl)-1,2,3,4-tetrahydroisoquinoline hydrochloride
Formel:
C₁₀H₁₀F₃N·ClH Molecular Weight: 237,649 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD05861559 CAS-Nummer: 1074764-70-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...