2-Chloro-6-fluoro-1H-benzo[d]imidazole 96%

Lieferant: Apollo Scientific

PC201022-250MG
APOSPC201022-250MGEA 74.2 EUR
APOSPC201022-250MG
2-Chloro-6-fluoro-1H-benzo[d]imidazole 96%
2-Chloro-6-fluoro-1H-benzo[d]imidazole
Formel: C₇H₄ClFN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD08457967
CAS-Nummer: 108662-49-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR