1-Chlor-2,4-difluor-3-nitrobenzol 97%

Lieferant: Apollo Scientific
Achtung

PC201032-1G PC201032-250MG PC201032-5G
APOSPC201032-1GEA 50.9 EUR
APOSPC201032-1G APOSPC201032-250MG APOSPC201032-5G
1-Chlor-2,4-difluor-3-nitrobenzol 97%
1-Chloro-2,4-difluoro-3-nitrobenzene
Formel: C₆H₂ClF₂NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD10686546
CAS-Nummer: 1151767-58-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR