6-Chloro-4-(trifluoromethyl)picolinonitrile 97%

Lieferant: Apollo Scientific

PC201034-1G PC201034-250MG
APOSPC201034-1GEA 763 EUR
APOSPC201034-1G APOSPC201034-250MG
6-Chloro-4-(trifluoromethyl)picolinonitrile 97%
6-Chloro-4-(trifluoromethyl)picolinonitrile
Formel: C₇H₂ClF₃N₂
Molecular Weight: 206.552 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00106773
CAS-Nummer: 1156542-25-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR