(E)-1-(2-Nitrovinyl)-3-(trifluoromethyl)benzene 97%

Lieferant: Apollo Scientific

PC201035-1G
APOSPC201035-1GEA 88.6 EUR
APOSPC201035-1G
(E)-1-(2-Nitrovinyl)-3-(trifluoromethyl)benzene 97%
(E)-1-(2-Nitrovinyl)-3-(trifluoromethyl)benzene
Formel: C₉H₆F₃NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00176733
CAS-Nummer: 115665-96-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR