6-(Trifluoromethyl)-1H-pyrrolo[3,2-b]pyridine 97%

Lieferant: Apollo Scientific

PC201043-100MG PC201043-1G PC201043-250MG
APOSPC201043-100MGEA 114 EUR
APOSPC201043-100MG APOSPC201043-1G APOSPC201043-250MG
6-(Trifluoromethyl)-1H-pyrrolo[3,2-b]pyridine 97%
6-(Trifluoromethyl)-1H-pyrrolo[3,2-b]pyridine
Formel: C₈H₅F₃N₂
Molecular Weight: 186.134 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12963187
CAS-Nummer: 1190311-44-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR