(6-(Difluoromethoxy)pyridin-3-yl)methanamine 97%

Lieferant: Apollo Scientific

PC201048-100MG
APOSPC201048-100MGEA 165 EUR
APOSPC201048-100MG
(6-(Difluoromethoxy)pyridin-3-yl)methanamine 97%
(6-(Difluoromethoxy)pyridin-3-yl)methanamine
Formel: C₇H₈F₂N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD18259135
CAS-Nummer: 1198103-43-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR