4-Bromo-2-chloro-3-fluoropyridine 95+%
Lieferant: Apollo Scientific
PC201060-100MG
PC201060-1G
PC201060-250MG
APOSPC201060-100MGEA
105
EUR
APOSPC201060-100MG
APOSPC201060-1G
APOSPC201060-250MG
4-Bromo-2-chloro-3-fluoropyridine 95+%
4-Bromo-2-chloro-3-fluoropyridine
Formel:
C₅H₂BrClFN Molecular Weight: 210.431 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD16606306 CAS-Nummer: 1211526-56-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...