4-Bromo-2-chloro-3-fluoropyridine 95+%

Lieferant: Apollo Scientific

PC201060-100MG PC201060-1G PC201060-250MG
APOSPC201060-100MGEA 105 EUR
APOSPC201060-100MG APOSPC201060-1G APOSPC201060-250MG
4-Bromo-2-chloro-3-fluoropyridine 95+%
4-Bromo-2-chloro-3-fluoropyridine
Formel: C₅H₂BrClFN
Molecular Weight: 210.431 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16606306
CAS-Nummer: 1211526-56-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR