Ethyl 2-bromo-6-fluorobenzoate 98%

Lieferant: Apollo Scientific

PC201076-1G
APOSPC201076-1GEA 68.3 EUR
APOSPC201076-1G
Ethyl 2-bromo-6-fluorobenzoate 98%
Ethyl 2-bromo-6-fluorobenzoate
Formel: C₉H₈BrFO₂
Molecular Weight: 247,061 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10000924
CAS-Nummer: 1214362-62-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR