6-Bromo-4-fluoro-1H-indazol-3-amine 97%

Lieferant: Apollo Scientific

PC201082-1G
APOSPC201082-1GEA 148 EUR
APOSPC201082-1G
6-Bromo-4-fluoro-1H-indazol-3-amine 97%
6-Bromo-4-fluoro-1H-indazol-3-amine
Formel: C₇H₅BrFN₃
Molecular Weight: 230.037 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD24368896
CAS-Nummer: 1227912-19-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR