(5-Fluoropyridin-2-yl)methanamine hydrochloride 95+%
Lieferant: Apollo Scientific
PC201084-250MG
APOSPC201084-250MGEA
143
EUR
APOSPC201084-250MG
(5-Fluoropyridin-2-yl)methanamine hydrochloride 95+%
(5-Fluoropyridin-2-yl)methanamine hydrochloride
Formel:
C₆H₇FN₂·ClH Lagertemperatur: Raumtemperatur |
MDL:
MFCD11505886 CAS-Nummer: 1228788-32-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...