1-(2-(Trifluoromethyl)phenyl)cyclopropanamine hydrochloride 95+%

Lieferant: Apollo Scientific

PC201085-100MG PC201085-1G PC201085-250MG
APOSPC201085-100MGEA 42.2 EUR
APOSPC201085-100MG APOSPC201085-1G APOSPC201085-250MG
1-(2-(Trifluoromethyl)phenyl)cyclopropanamine hydrochloride 95+%
1-(2-(Trifluoromethyl)phenyl)cyclopropanamine hydrochloride
Formel: C₁₀H₁₀F₃N·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD09028131
CAS-Nummer: 1228879-24-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR