1-(2-(Trifluoromethyl)phenyl)cyclopropanamine hydrochloride 95+%
Lieferant: Apollo Scientific
PC201085-100MG
PC201085-1G
PC201085-250MG
APOSPC201085-100MGEA
42.2
EUR
APOSPC201085-100MG
APOSPC201085-1G
APOSPC201085-250MG
1-(2-(Trifluoromethyl)phenyl)cyclopropanamine hydrochloride 95+%
1-(2-(Trifluoromethyl)phenyl)cyclopropanamine hydrochloride
Formel:
C₁₀H₁₀F₃N·ClH Lagertemperatur: Raumtemperatur |
MDL:
MFCD09028131 CAS-Nummer: 1228879-24-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...