2-Ethoxy-6-(trifluoromethyl)pyridine 95+%

Lieferant: Apollo Scientific
Achtung

PC201090-1G
APOSPC201090-1GEA 179 EUR
APOSPC201090-1G
2-Ethoxy-6-(trifluoromethyl)pyridine 95+%
2-Ethoxy-6-(trifluoromethyl)pyridine
Formel: C₈H₈F₃NO
Molecular Weight: 191,15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD17015840
CAS-Nummer: 1245563-18-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR