Methyl-4-fluoro-1H-indole-5-carboxylate 95%

Lieferant: Apollo Scientific

Synonyme: Methyl 4-fluoro-1H-indole-5-carboxylate

PC201093-100MG PC201093-1G PC201093-250MG
APOSPC201093-100MGEA 249 EUR
APOSPC201093-100MG APOSPC201093-1G APOSPC201093-250MG
Methyl-4-fluoro-1H-indole-5-carboxylate 95%
Methyl-4-fluoro-1H-indole-5-carboxylate
Formel: C₁₀H₈FNO₂
Molecular Weight: 193.17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD21336690
CAS-Nummer: 1252782-43-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR