Methyl 4-(chloromethyl)-3,5-difluorobenzoate 97%

Lieferant: Apollo Scientific

PC201105-250MG
APOSPC201105-250MGEA 80 EUR
APOSPC201105-250MG
Methyl 4-(chloromethyl)-3,5-difluorobenzoate 97%
Methyl 4-(chloromethyl)-3,5-difluorobenzoate
Formel: C₉H₇ClF₂O₂
Molecular Weight: 220,6 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD17676601
CAS-Nummer: 1263283-69-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR