(6-(Trifluoromethyl)pyridin-2-yl)methanol 98%

Lieferant: Apollo Scientific

PC201117-1G
APOSPC201117-1GEA 56.7 EUR
APOSPC201117-1G
(6-(Trifluoromethyl)pyridin-2-yl)methanol 98%
(6-(Trifluoromethyl)pyridin-2-yl)methanol
Formel: C₇H₆F₃NO
Molecular Weight: 177,124 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11100723
CAS-Nummer: 131747-53-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR