trans-4-(Trifluoromethyl)cyclohexanecarboxylic acid 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC201120-5G
APOSPC201120-5GEA
68.3
EUR
APOSPC201120-5G
trans-4-(Trifluoromethyl)cyclohexanecarboxylic acid 98%
trans-4-(Trifluoromethyl)cyclohexanecarboxylic acid
Formel:
C₈H₁₁F₃O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD09037363 CAS-Nummer: 133261-33-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...