4-Bromo-5-fluoro-2-methylbenzoic acid 98%

Lieferant: Apollo Scientific

PC201125-5G
APOSPC201125-5GEA 108 EUR
APOSPC201125-5G
4-Bromo-5-fluoro-2-methylbenzoic acid 98%
4-Bromo-5-fluoro-2-methylbenzoic acid
Formel: C₈H₆BrFO₂
Molecular Weight: 233.034 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18917157
CAS-Nummer: 1349715-55-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR