Ethyl 4-amino-3,5-difluorobenzoate 95+%
Lieferant: Apollo Scientific
|
Achtung
|
PC201137-1G
APOSPC201137-1GEA
104
EUR
APOSPC201137-1G
Ethyl 4-amino-3,5-difluorobenzoate 95+%
Ethyl 4-amino-3,5-difluorobenzoate
Formel:
C₉H₉F₂NO₂ Molecular Weight: 201,17 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD28678682 CAS-Nummer: 1415920-00-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...