5-(Trifluoromethyl)indoline 97+%

Lieferant: Apollo Scientific

PC201160-250MG
APOSPC201160-250MGEA 93.1 EUR
APOSPC201160-250MG
5-(Trifluoromethyl)indoline 97+%
5-(Trifluoromethyl)indoline
Formel: C₉H₈F₃N
Molecular Weight: 187,162 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03095322
CAS-Nummer: 162100-55-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR