5-(Aminomethyl)-2-fluorobenzonitrile hydrochloride 97%

Lieferant: Apollo Scientific

PC201161-250MG
APOSPC201161-250MGEA 98.9 EUR
APOSPC201161-250MG
5-(Aminomethyl)-2-fluorobenzonitrile hydrochloride 97%
5-(Aminomethyl)-2-fluorobenzonitrile hydrochloride
Formel: C₈H₇FN₂·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD28125524
CAS-Nummer: 1638487-42-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR