2-Chloro-N-(2,2,2-trifluoroethyl)acetamide 98%

Lieferant: Apollo Scientific

PC201170-250MG
APOSPC201170-250MGEA 78.3 EUR
APOSPC201170-250MG
2-Chloro-N-(2,2,2-trifluoroethyl)acetamide 98%
2-Chloro-N-(2,2,2-trifluoroethyl)acetamide
Formel: C₄H₅ClF₃NO
Molecular Weight: 175,537 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03984449
CAS-Nummer: 170655-44-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR