3-Brom-4-(trifluormethyl)anilin

Lieferant: Apollo Scientific
Gefahr

PC201171-5G PC201171-25G
APOSPC201171-5GEA 81.2 EUR
APOSPC201171-5G APOSPC201171-25G
3-Brom-4-(trifluormethyl)anilin
3-Brom-4-(trifluormethyl)anilin
Formel: C₇H₅BrF₃N
Lagertemperatur: Raumtemperatur
MDL: MFCD12828280
CAS-Nummer: 172215-91-7
UN: 2810
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR