4-Ethoxy-2-fluoropyridine 97%

Lieferant: Apollo Scientific

PC201174-100MG PC201174-1G PC201174-250MG
APOSPC201174-100MGEA 27.7 EUR
APOSPC201174-100MG APOSPC201174-1G APOSPC201174-250MG
4-Ethoxy-2-fluoropyridine 97%
4-Ethoxy-2-fluoropyridine
Formel: C₇H₈FNO
Lagertemperatur: Raumtemperatur
MDL: MFCD09835274
CAS-Nummer: 175965-82-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR