1-Chloro-2,5-difluoro-3-nitrobenzene 95+%
Lieferant: Apollo Scientific
PC201180-100MG
PC201180-1G
PC201180-250MG
APOSPC201180-100MGEA
49.3
EUR
APOSPC201180-100MG
APOSPC201180-1G
APOSPC201180-250MG
1-Chloro-2,5-difluoro-3-nitrobenzene 95+%
1-Chloro-2,5-difluoro-3-nitrobenzene
Formel:
C₆H₂ClF₂NO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD27923474 CAS-Nummer: 1805029-24-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...